siRNA Details
Gene Symbol: BC025920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030677.1 | 3 | 1329 | |||
GenBank mRNA | AK140772.1 | - | 3 | 1330 | ||
AK142150.1 | - | 1 | 2251 | |||
BC025920.1 | - | 2 | 2009 | |||
BC145724.1 | - | 2 | 1312 | |||
BC145726.1 | - | 2 | 1341 |
