siRNA Details
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028417.1 | 2 | 453 | |||
GenBank mRNA | AK041797.1 | - | 2 | 454 | ||
AK138349.1 | - | 2 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB638505.1 | - | 2 | 454 | Chr. 5 - 43190260 - 43235354 [-] on Build GRCm38 |
