siRNA Details
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028417.1 | 3, 4 | 580 | |||
GenBank mRNA | AK041797.1 | - | 3, 4 | 581 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm7854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028417.1 | 3, 4 | 580 | |||
GenBank mRNA | AK041797.1 | - | 3, 4 | 581 |