siRNA Details
Gene Symbol: 4930592I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033307.1 | 3 | 356 | |||
GenBank mRNA | AK077183.1 | - | 3 | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617118.1 | - | 3 | 345 | Chr. 18 - 82918739 - 82920548 [-] on Build GRCm38 |
