siRNA Details
Gene Symbol: 4930579K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029444.1 | 1 | 497 | |||
GenBank mRNA | AK016329.1 | - | 1 | 469 | ||
AK077474.1 | - | 1 | 479 | |||
AK087620.1 | - | 1 | 498 | |||
BC092140.1 | - | 1 | 438 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715968.1 | - | 1 | 469 | Chr. 9 - 98562678 - 98563583 [-] on Build GRCm38 |
