siRNA Details
Gene Symbol: 1700020N01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027968.1 | 3 | 557 | |||
GenBank mRNA | AK006179.1 | - | 3 | 545 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706276.1 | - | 3 | 545 | Chr. 10 - 21593157 - 21622021 [+] on Build GRCm38 |
