siRNA Details
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002840.2 | 1 | 1078 | |||
GenBank mRNA | AK083453.1 | - | 1 | 1079 | ||
BC080708.1 | - | 2 | 325 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gas5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002840.2 | 1 | 1078 | |||
GenBank mRNA | AK083453.1 | - | 1 | 1079 | ||
BC080708.1 | - | 2 | 325 |