siRNA Details
Gene Symbol: 1810058I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015608.1 | 3 | 569 | |||
NR_027875.1 | 3 | 799 | ||||
GenBank mRNA | AK004512.1 | - | 3 | 528 | ||
AK007895.1 | - | 3 | 559 | |||
AK082166.1 | - | 615 | ||||
AK155176.1 | - | 3 | 559 | |||
AK158055.1 | - | 3 | 800 | |||
AK171831.1 | - | 3 | 569 | |||
CA751247.1 | - | 1 | 132 |
