siRNA Details
Gene Symbol: 4932414J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028259.1 | 9 | 2949 | |||
GenBank mRNA | AK030001.1 | - | 9 | 2950 |
Gene Symbol: 4932415M13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073205.1 | 8 | 2831 | |||
GenBank mRNA | AK030017.1 | - | 8 | 2832 |
