siRNA Details
Gene Symbol: Gm17769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027377.1 | 1 | 452 | |||
GenBank mRNA | AK141975.1 | - | 1 | 453 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm17769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027377.1 | 1 | 452 | |||
GenBank mRNA | AK141975.1 | - | 1 | 453 |