siRNA Details
Gene Symbol: 4930528A17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028384.1 | 1 | 1620 | |||
GenBank mRNA | AK015922.1 | - | 1 | 1542 | ||
AK039347.1 | - | 1 | 1604 | |||
BC031206.1 | - | 1 | 1585 |
