siRNA Details
Gene Symbol: 4930583K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027879.1 | 2 | 554 | |||
GenBank mRNA | AK016343.1 | - | 2 | 555 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930583K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027879.1 | 2 | 554 | |||
GenBank mRNA | AK016343.1 | - | 2 | 555 |