siRNA Details
Gene Symbol: 4930584F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029445.1 | 2 | 165 | |||
GenBank mRNA | AK016349.1 | - | 2 | 132 | ||
BC049642.1 | - | 2 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU935604.1 | - | 2 | 143 | Chr. 5 - 26479718 - 26493289 [-] on Build GRCm38 |
