siRNA Details
Gene Symbol: 4930584F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029445.1 | 3 | 679 | |||
GenBank mRNA | AK005600.1 | - | 4 | 612 | ||
AK015867.1 | - | 2 | 377 | |||
AK016349.1 | - | 3 | 424 | |||
BC049642.1 | - | 3 | 654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715418.1 | - | 2 | 377 | Chr. 5 - 26474935 - 26493289 [-] on Build GRCm38 |
