siRNA Details
Gene Symbol: Bach2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026843.1 | 4 | 484 | |||
GenBank mRNA | AK144923.1 | - | 4 | 1462 | ||
BC024582.1 | - | 4 | 484 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI150671.1 | - | 4 | 485 | Chr. 4 - 32561587 - 32571662 [-] on Build GRCm38 |
