siRNA Details
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015561.2 | 4 | 1088 | |||
GenBank mRNA | AK013308.1 | - | 2 | 994 | ||
AK039679.1 | - | 4 | 1089 | |||
AK047851.1 | - | 260 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015561.2 | 4 | 1088 | |||
GenBank mRNA | AK013308.1 | - | 2 | 994 | ||
AK039679.1 | - | 4 | 1089 | |||
AK047851.1 | - | 260 |