siRNA Details
Gene Symbol: BC006965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024085.1 | 2 | 380 | |||
GenBank mRNA | AK035132.1 | - | 448 | |||
AK048256.1 | - | 2 | 381 | |||
AK145007.1 | - | 2 | 383 | |||
BC006965.1 | - | 3 | 554 | |||
BC010247.1 | - | 3 | 272 |
