siRNA Details
Gene Symbol: BC006965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024085.1 | 1 | 154 | |||
GenBank mRNA | AK048256.1 | - | 1 | 155 | ||
AK145007.1 | - | 1 | 157 | |||
AK157980.1 | - | 1 | 161 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC006965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024085.1 | 1 | 154 | |||
GenBank mRNA | AK048256.1 | - | 1 | 155 | ||
AK145007.1 | - | 1 | 157 | |||
AK157980.1 | - | 1 | 161 |