siRNA Details
Gene Symbol: 6530411M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027881.1 | 2 | 752 | |||
GenBank mRNA | AK078247.1 | - | 2 | 567 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY718979.1 | - | 2 | 567 | Chr. 17 - 9149174 - 9167836 [-] on Build GRCm38 |
