siRNA Details
Gene Symbol: Speer9-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001583.3 | 3 | 383 | |||
GenBank mRNA | BY714690.1 | - | 3 | 384 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK015142.1 | - | 384 | Chr. - - [-] on Build |
