siRNA Details
Gene Symbol: LOC106740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027905.1 | 1 | 749 | |||
GenBank mRNA | AK134370.1 | - | 1 | 749 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV024517.2 | - | 1 | 574 | Chr. 17 - 14947738 - 14948648 [-] on Build GRCm38 |
