siRNA Details
Gene Symbol: Kis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_003188.1 | 1 | 798 | |||
GenBank mRNA | AK084356.1 | - | 2 | 230 | ||
AY940614.1 | - | LIT3387 Kis2 |
1 | 798 | ||
AY940615.1 | - | LIT3388 Kis2 |
2 | 484 | ||
AY940616.1 | - | LIT3389 Kis2 |
1 | 117 |
