siRNA Details
Gene Symbol: 8430431K14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002849.1 | 1 | 2548 | |||
GenBank mRNA | AK046503.1 | - | 1 | 2548 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB321574.2 | - | 1 | 400 | Chr. 19 - 31213543 - 31214248 [-] on Build GRCm38 |
