siRNA Details
Gene Symbol: C030034I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026848.1 | 1 | 387 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE949423.1 | - | 1 | 407 | Chr. 17 - 69416427 - 69416975 [+] on Build GRCm38 |
