siRNA Details
Gene Symbol: 5830416P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028427.1 | 5 | 2056 | |||
GenBank mRNA | AK158034.1 | - | 5 | 2045 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5830416P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028427.1 | 5 | 2056 | |||
GenBank mRNA | AK158034.1 | - | 5 | 2045 |