siRNA Details
Gene Symbol: 5830416P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028427.1 | 2 | 123 | |||
GenBank mRNA | AK017935.1 | - | 2 | 124 | ||
AK158034.1 | - | 2 | 111 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY752794.1 | - | 2 | 108 | Chr. 19 - 53451001 - 53464781 [-] on Build GRCm38 |
