siRNA Details
Gene Symbol: D930016D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030673.1 | 4 | 1850 | |||
GenBank mRNA | AK086246.1 | - | 1 | 1609 | ||
AK140900.1 | - | 2 | 1405 | |||
BC060616.1 | - | 4 | 1817 | |||
BC082597.1 | - | 2326 |
