siRNA Details
Gene Symbol: D930016D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030673.1 | 4 | 1821 | |||
GenBank mRNA | AK086246.1 | - | 1 | 1580 | ||
AK140900.1 | - | 2 | 1376 | |||
BC023132.1 | - | 1 | 432 | |||
BC060616.1 | - | 4 | 1788 | |||
BC082597.1 | - | 2297 |
