siRNA Details
Gene Symbol: D930016D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030673.1 | 4 | 2513 | |||
GenBank mRNA | AK081399.1 | - | 1 | 206 | ||
AK140900.1 | - | 2 | 2068 | |||
BC023132.1 | - | 1 | 1123 | |||
BC060616.1 | - | 4 | 2480 | |||
BC082597.1 | - | 2989 |
