siRNA Details
Gene Symbol: 2810025M15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027984.1 | 3 | 1091 | |||
GenBank mRNA | AK012812.1 | - | 3 | 667 | ||
BC055845.1 | - | 3 | 1091 | 3585495 | ||
BC055878.1 | - | 3 | 677 | 4222031 |
