siRNA Details
Gene Symbol: 2810403D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015493.2 | 2 | 237 | |||
GenBank mRNA | AK007586.1 | - | 2 | 146 | ||
AK012972.1 | - | 2 | 162 | |||
AK039349.1 | - | 1 | 211 | |||
AK086455.1 | - | 2 | 238 | |||
AK155344.1 | - | 2 | 140 |
