siRNA Details
Gene Symbol: 2810403D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015493.2 | 5 | 680 | |||
GenBank mRNA | AK007586.1 | - | 3 | 434 | ||
AK086455.1 | - | 5 | 681 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707631.1 | - | 3 | 428 | Chr. X - 108834665 - 108886683 [+] on Build GRCm38 |
