siRNA Details
Gene Symbol: 2810403D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015493.2 | 3 | 330 | |||
GenBank mRNA | AK019485.1 | - | 1 | 2332 | ||
AK086455.1 | - | 3 | 331 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB663071.1 | - | 3 | 331 | Chr. X - 108834478 - 108886514 [+] on Build GRCm38 |
