siRNA Details
Gene Symbol: 1700096J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027883.1 | 2 | 389 | |||
GenBank mRNA | AK006164.1 | - | 2 | 390 | ||
AK007092.1 | - | 2 | 376 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706265.1 | - | 2 | 390 | Chr. 11 - 109346867 - 109353603 [+] on Build GRCm38 |
