siRNA Details
Gene Symbol: 6230400D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029446.1 | 1 | 646 | |||
GenBank mRNA | AK078695.1 | - | 1 | 579 | ||
AK156493.1 | - | 558 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 6230400D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029446.1 | 1 | 646 | |||
GenBank mRNA | AK078695.1 | - | 1 | 579 | ||
AK156493.1 | - | 558 |