siRNA Details
Gene Symbol: 6230400D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029446.1 | 1 | 610 | |||
GenBank mRNA | AK078695.1 | - | 1 | 543 | ||
AK156493.1 | - | 522 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB064773.2 | - | 1 | 201 | Chr. 14 - 20702012 - 20702681 [-] on Build GRCm38 |
