siRNA Details
Gene Symbol: Gm4956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002858.1 | 6 | 935 | |||
GenBank mRNA | BC061226.1 | - | 6 | 935 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002858.1 | 6 | 935 | |||
GenBank mRNA | BC061226.1 | - | 6 | 935 |