siRNA Details
Gene Symbol: Copg2os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002845.1 | 1461 | ||||
GenBank mRNA | AK045748.1 | - | 687 | |||
U20264.1 | - | 1461 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Copg2os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002845.1 | 1461 | ||||
GenBank mRNA | AK045748.1 | - | 687 | |||
U20264.1 | - | 1461 |