siRNA Details
Gene Symbol: E030011O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015511.2 | 3 | 565 | |||
GenBank mRNA | AK086536.1 | - | 3 | 566 | ||
AK136643.1 | - | 422 |

More Information
Other Species Targeted

Gene Ontology Categories:
