siRNA Details
Gene Symbol: E030011O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015511.2 | 1 | 231 | |||
GenBank mRNA | AK053135.1 | - | 1 | 128 | ||
AK086536.1 | - | 1 | 232 | |||
AK136643.1 | - | 88 | ||||
AK211984.1 | - | 1 | 44 |
