siRNA Details
Gene Symbol: E030011O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015511.2 | 1 | 381 | |||
GenBank mRNA | AK053135.1 | - | 1 | 278 | ||
AK086536.1 | - | 1 | 382 | |||
AK136643.1 | - | 238 | ||||
AK194500.1 | - | 1 | 63 |
