siRNA Details
Gene Symbol: E030011O05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015511.2 | 1, 2 | 454 | |||
GenBank mRNA | AK086536.1 | - | 1, 2 | 455 | ||
AK136643.1 | - | 311 | ||||
AK194500.1 | - | 1, 2 | 136 | |||
AK202778.1 | - | 1, 2 | 136 |
