siRNA Details
Gene Symbol: 1700028J19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029436.1 | 5 | 680 | |||
GenBank mRNA | AK006462.1 | - | 5 | 742 | ||
BC049653.1 | - | 5 | 646 | |||
BC061239.1 | - | 4 | 556 | |||
BC087938.1 | - | 4 | 556 | |||
BC100477.1 | - | 5 | 662 |
