siRNA Details
Gene Symbol: E130114P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015513.2 | 3 | 509 | |||
GenBank mRNA | AK053617.1 | - | 669 | |||
AK082601.1 | - | 3 | 511 | |||
AK086199.1 | - | 845 | ||||
AK086602.1 | - | 3 | 555 | |||
BC048947.1 | - | 4 | 628 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177023.1 | - | 845 | Chr. - - [-] on Build |
