siRNA Details
Gene Symbol: 5330413P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029381.1 | 3 | 572 | |||
GenBank mRNA | AK030453.1 | - | 3 | 571 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617812.1 | - | 3 | 571 | Chr. 2 - 131822951 - 131847655 [-] on Build GRCm38 |
