siRNA Details
Gene Symbol: 9330151L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033222.1 | 2 | 2485 | |||
GenBank mRNA | AK034052.1 | - | 2 | 2473 | ||
AK154174.1 | - | 2 | 2485 | |||
AK161156.1 | - | 1 | 2602 |
