siRNA Details
Gene Symbol: Gm10012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028042.1 | 1 | 157 | |||
GenBank mRNA | CA533111.1 | - | 1 | 157 |
Gene Symbol: Gm12338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137335.1 | - | 1418 |
