siRNA Details
Gene Symbol: 1700049L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003644.1 | 1 | 431 | |||
GenBank mRNA | AK075762.1 | - | 1 | 436 | ||
BC048504.1 | - | 1 | 431 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV045951.3 | - | 1 | 628 | Chr. 10 - 71979886 - 71980698 [-] on Build GRCm38 |
