siRNA Details
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029459.1 | 3 | 645 | |||
GenBank mRNA | BC055849.1 | - | 3 | 565 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM942436.1 | - | 3 | 526 | Chr. 11 - 117115291 - 117128337 [+] on Build GRCm38 |
