siRNA Details
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029459.1 | 3 | 551 | |||
GenBank mRNA | BC055849.1 | - | 3 | 471 | ||
CJ066501.1 | - | 3 | 489 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM942436.1 | - | 3 | 432 | Chr. 11 - 117115291 - 117128337 [+] on Build GRCm38 |
